Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032837.1 | NP_001028009.1 | 344 | 88 | ||
GenBank mRNA | AF322912.1 | - | 344 | 88 | ||
JU475599.1 | - | 680 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |