Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD40LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032839.1 | NP_001028011.1 | 188 | 122 | ||
GenBank mRNA | AF344859.1 | - | 188 | 122 | ||
JU471432.1 | - | 207 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta Inventoried |