Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD40LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032839.1 | NP_001028011.1 | 326 | 121 | ||
GenBank mRNA | AF344859.1 | - | 326 | 121 | ||
JU471432.1 | - | 345 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |