Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINLW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032841.1 | NP_001028013.1 | 157 | 81 | ||
GenBank mRNA | AF346414.1 | - | 157 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |