Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032939.1 | NP_001028111.1 | 262 | 80 | ||
GenBank mRNA | AF366397.1 | - | 262 | 80 | ||
JU336136.1 | - | 379 | 80 | |||
JU476998.1 | - | 381 | 80 | |||
JV660075.1 | - | 403 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |