Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032956.1 | NP_001028128.1 | 243 | 88 | ||
GenBank mRNA | AF366399.1 | - | 99 | 88 | ||
AY821556.1 | - | 243 | 88 | |||
JU336137.1 | - | 478 | 88 | |||
JV694854.1 | - | 504 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |