Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032956.1 | NP_001028128.1 | 480 | 68 | ||
GenBank mRNA | AF366399.1 | - | 336 | 68 | ||
AY821556.1 | - | 480 | 68 | |||
JU336137.1 | - | 715 | 68 | |||
JV694854.1 | - | 741 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |