Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032961.1 | NP_001028133.1 | 191 | 95 | ||
XM_015150994.2 | XP_015006480.1 | 686 | 95 | |||
GenBank mRNA | AJ622828.1 | - | 191 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |