Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032902.1 | NP_001028074.1 | 471 | 75 | ||
GenBank mRNA | AY350447.1 | - | 471 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Macaca mulatta |