Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032903.1 | NP_001028075.1 | 228 | 88 | ||
GenBank mRNA | AY371699.1 | - | 228 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |