Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039950.1 | NP_001035039.1 | 356 | 68 | ||
GenBank mRNA | AY386684.1 | - | 356 | 68 | ||
AY386688.1 | - | 356 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |