Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02788869_m1
See other ADGRG1 GE Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
EGK_12829, GPR56
Chromosome Location:
Chr.20: 44855566 - 44904737 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02788869_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ADGRG1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001033020.1
NP_001028192.1
1051
67
XM_015126156.2
XP_014981642.1
1199
67
XM_015126157.2
XP_014981643.1
1345
67
XM_015126158.2
XP_014981644.1
1157
67
XM_015126161.2
XP_014981647.1
1157
67
XM_015126162.2
XP_014981648.1
1175
67
XM_015126163.2
XP_014981649.1
1227
67
XM_015126164.2
XP_014981650.1
1196
67
XM_015126165.2
XP_014981651.1
1226
67
XM_015126166.2
XP_014981652.1
1395
67
XM_015126167.2
XP_014981653.1
1142
67
XM_015126168.2
XP_014981654.1
1187
67
XM_015126169.2
XP_014981655.1
1208
67
XM_015126170.2
XP_014981656.1
1159
67
XM_015126171.2
XP_014981657.1
1644
67
XM_015126172.2
XP_014981658.1
1629
67
XM_028840341.1
XP_028696174.1
1090
67
XM_028840342.1
XP_028696175.1
1087
67
XM_028840344.1
XP_028696177.1
1292
67
XM_028840345.1
XP_028696178.1
1074
67
XM_028840346.1
XP_028696179.1
1099
67
XM_028840347.1
XP_028696180.1
1149
67
GenBank mRNA
AY845352.1
-
1051
67
JU323293.1
-
1285
67
JU337110.1
-
1285
67
JU470897.1
-
1295
67
JU470898.1
-
1280
67
JU477855.1
-
1280
67
JV601061.1
-
1285
67
JV635411.1
-
1259
67
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
G-protein coupled receptor
Macaca mulatta
Panther Classification:
Molecular Function -
G-protein coupled receptor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0