Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195425.1 | NP_001182354.1 | 251 | 65 | ||
XM_015116593.2 | XP_014972079.1 | 364 | 65 | |||
XM_028851407.1 | XP_028707240.1 | 364 | 65 | |||
GenBank mRNA | AY964932.1 | - | 131 | 65 | ||
CN642704.1 | - | 514 | 65 | |||
CO726805.1 | - | 290 | 65 | |||
JV059632.1 | - | 267 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |