Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195502.1 | NP_001182431.1 | 269 | 73 | ||
GenBank mRNA | CD767990.1 | - | 269 | 73 | ||
DQ148153.1 | - | 257 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |