Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042433.2 | NP_001035898.1 | 2003 | 72 | ||
XM_015121466.2 | XP_014976952.1 | 2313 | 72 | |||
GenBank mRNA | DQ644021.1 | - | 2006 | 72 | ||
EF467993.1 | - | 2041 | 72 | |||
EF467994.1 | - | 2041 | 72 | |||
JU470652.1 | - | 2210 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |