Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042787.1 | NP_001036252.1 | 623 | 104 | ||
XM_015147030.2 | XP_015002516.2 | 966 | 104 | |||
GenBank mRNA | U87259.1 | - | 623 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |