Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040211.1 | NP_001035301.1 | 1150 | 138 | ||
GenBank mRNA | AY635463.1 | - | 1150 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |