Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040671.1 | NP_001035761.1 | 1946 | 118 | ||
GenBank mRNA | DQ155429.1 | - | 1946 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |