Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092818.4 | XP_001092818.1 | 812 | 143 | ||
GenBank mRNA | JU332625.1 | - | 779 | 143 | ||
JU474084.1 | - | 753 | 143 | |||
JV045994.1 | - | 764 | 143 | |||
JV604088.1 | - | 782 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |