Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258081.1 | NP_001245010.1 | 1161 | 94 | ||
GenBank mRNA | JU473276.1 | - | 1161 | 94 | ||
JV044222.1 | - | 1324 | 94 | |||
JV533763.1 | - | 1398 | 94 | |||
JV571640.1 | - | 1226 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |