Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261675.1 | NP_001248604.1 | 523 | 131 | ||
XM_015135551.2 | XP_014991037.1 | 714 | 131 | |||
GenBank mRNA | JU334882.1 | - | 530 | 131 | ||
JV047299.1 | - | 523 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |