Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260609.2 | NP_001247538.1 | 733 | 120 | ||
XM_015146783.2 | XP_015002269.1 | 695 | 120 | |||
GenBank mRNA | JU335513.1 | - | 728 | 120 | ||
JU476453.1 | - | 734 | 120 | |||
JV047692.1 | - | 733 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |