Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261368.1 | NP_001248297.1 | 131 | 72 | ||
GenBank mRNA | JU322518.1 | - | 93 | 72 | ||
JU473306.1 | - | 157 | 72 | |||
JV045674.1 | - | 131 | 72 | |||
JV601919.1 | - | 175 | 72 |