Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261583.1 | NP_001248512.1 | 40 | 51 | ||
GenBank mRNA | JU323205.1 | - | 40 | 51 | ||
JU474606.1 | - | 41 | 51 | |||
JV044715.1 | - | 40 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |