Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261583.1 | NP_001248512.1 | 618 | 102 | ||
XM_015135409.2 | XP_014990895.1 | 828 | 102 | |||
XM_028846756.1 | XP_028702589.1 | 837 | 102 | |||
GenBank mRNA | JU323205.1 | - | 618 | 102 | ||
JU474606.1 | - | 619 | 102 | |||
JV044715.1 | - | 618 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |