Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260791.1 | NP_001247720.1 | 415 | 70 | ||
GenBank mRNA | JU477891.1 | - | 415 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |