Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266635.1 | NP_001253564.1 | 261 | 86 | ||
XM_028831501.1 | XP_028687334.1 | 315 | 86 | |||
GenBank mRNA | JU332417.1 | - | 220 | 86 | ||
JU472598.1 | - | 189 | 86 | |||
JV043962.1 | - | 261 | 86 | |||
JV602978.1 | - | 289 | 86 | |||
JV711112.1 | - | 2685 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |