Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266970.1 | NP_001253899.1 | 639 | 84 | ||
XM_015130224.2 | XP_014985710.1 | 902 | 84 | |||
XM_015130228.2 | XP_014985714.2 | 1276 | 84 | |||
XM_028843137.1 | XP_028698970.1 | 1218 | 84 | |||
XM_028843138.1 | XP_028698971.1 | 1146 | 84 | |||
XM_028843139.1 | XP_028698972.1 | 1119 | 84 | |||
GenBank mRNA | JU335870.1 | - | 627 | 84 | ||
JU476748.1 | - | 655 | 84 | |||
JV047902.1 | - | 639 | 84 | |||
JV625415.1 | - | 657 | 84 | |||
JV658509.1 | - | 648 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |