Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260912.1 | NP_001247841.1 | 1070 | 65 | ||
GenBank mRNA | JU321043.1 | - | 1280 | 65 | ||
JU476460.1 | - | 1256 | 65 | |||
JV043848.1 | - | 1070 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |