Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261580.1 | NP_001248509.1 | 521 | 128 | ||
GenBank mRNA | JU322637.1 | - | 521 | 128 | ||
KC734500.1 | - | 466 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |