Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193861.2 | NP_001180790.1 | 1277 | 100 | ||
XM_028852169.1 | XP_028708002.1 | 809 | 100 | |||
GenBank mRNA | JU325688.1 | - | 734 | 100 | ||
JV052988.1 | - | 734 | 100 | |||
JV643494.1 | - | 734 | 100 | |||
JV674297.1 | - | 732 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |