Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260599.1 | NP_001247528.1 | 868 | 119 | ||
GenBank mRNA | JV045264.1 | - | 868 | 119 | ||
JV045265.1 | - | 868 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |