Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260636.1 | NP_001247565.1 | 379 | 79 | ||
GenBank mRNA | JU322558.1 | - | 379 | 79 | ||
JV045703.1 | - | 372 | 79 | |||
JV690223.1 | - | 381 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |