Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083049.4 | XP_001083049.2 | 658 | 142 | ||
XM_015143179.2 | XP_014998665.2 | 161 | 142 | |||
XM_028851341.1 | XP_028707174.1 | 650 | 142 | |||
XM_028851342.1 | XP_028707175.1 | 650 | 142 | |||
GenBank mRNA | JU323393.1 | - | 641 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |