Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193728.2 | NP_001180657.2 | 375 | 123 | ||
GenBank mRNA | JU322596.1 | - | 301 | 123 | ||
JV532754.1 | - | 199 | 123 | |||
JV667268.1 | - | 287 | 123 | |||
JV703955.1 | - | 298 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |