Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148650.2 | XP_015004136.2 | 330 | 88 | ||
GenBank mRNA | JV527813.1 | - | 275 | 88 | ||
JV625116.1 | - | 242 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |