Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260709.1 | NP_001247638.1 | 1543 | 121 | ||
XM_015138312.2 | XP_014993798.1 | 1059 | 121 | |||
XM_028847961.1 | XP_028703794.1 | 1059 | 121 | |||
GenBank mRNA | JU321672.1 | - | 1548 | 121 | ||
JU321673.1 | - | 1548 | 121 | |||
JU471417.1 | - | 1543 | 121 | |||
JV048869.1 | - | 1481 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |