Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265663.2 | NP_001252592.1 | 536 | 120 | ||
GenBank mRNA | JV636279.1 | - | 538 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |