Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193749.1 | NP_001180678.1 | 493 | 101 | ||
GenBank mRNA | JU334140.1 | - | 330 | 101 | ||
JV046837.1 | - | 346 | 101 | |||
JV579453.1 | - | 362 | 101 | |||
JV649239.1 | - | 372 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |