Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265775.1 | NP_001252704.1 | 540 | 142 | ||
GenBank mRNA | JU327026.1 | - | 471 | 142 | ||
JV569723.1 | - | 540 | 142 | |||
JV667332.1 | - | 540 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |