Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193811.2 | NP_001180740.1 | 352 | 69 | ||
XM_015127083.2 | XP_014982569.1 | 379 | 69 | |||
XM_028841690.1 | XP_028697523.1 | 379 | 69 | |||
XM_028841691.1 | XP_028697524.1 | 379 | 69 | |||
GenBank mRNA | JU321136.1 | - | 285 | 69 | ||
JV045241.1 | - | 291 | 69 | |||
JV566744.1 | - | 271 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |