Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151742.2 | XP_015007228.2 | 609 | 65 | ||
XM_015151743.2 | XP_015007229.2 | 609 | 65 | |||
GenBank mRNA | JU344447.1 | - | 514 | 65 | ||
JU487895.1 | - | 492 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |