Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260608.1 | NP_001247537.1 | 426 | 115 | ||
GenBank mRNA | JU321610.1 | - | 492 | 115 | ||
JV045144.1 | - | 426 | 115 | |||
JV601372.1 | - | 434 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |