Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260608.1 | NP_001247537.1 | 754 | 127 | ||
XM_028849098.1 | XP_028704931.1 | 682 | 127 | |||
GenBank mRNA | JU321610.1 | - | 820 | 127 | ||
JV045144.1 | - | 754 | 127 | |||
JV601372.1 | - | 762 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |