Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266477.1 | NP_001253406.1 | 1300 | 105 | ||
XM_015145729.2 | XP_015001215.1 | 1167 | 105 | |||
GenBank mRNA | JU322448.1 | - | 1274 | 105 | ||
JU472205.1 | - | 1332 | 105 | |||
JV048087.1 | - | 1300 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |