Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266870.1 | NP_001253799.1 | 583 | 145 | ||
XM_028827272.1 | XP_028683105.1 | 1037 | 145 | |||
GenBank mRNA | JU334178.1 | - | 558 | 145 | ||
JU475311.1 | - | 556 | 145 | |||
JV046860.1 | - | 583 | 145 | |||
JV579683.1 | - | 650 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |