Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125785.2 | XP_014981271.1 | 415 | 105 | ||
XM_015125786.2 | XP_014981272.1 | 415 | 105 | |||
XM_015125788.2 | XP_014981274.1 | 415 | 105 | |||
XM_028841230.1 | XP_028697063.1 | 415 | 105 | |||
GenBank mRNA | JU334785.1 | - | 220 | 105 | ||
JU471813.1 | - | 354 | 105 | |||
JV047250.1 | - | 231 | 105 | |||
JV565039.1 | - | 322 | 105 | |||
JV600708.1 | - | 343 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |