Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261313.1 | NP_001248242.1 | 350 | 141 | ||
XM_028847996.1 | XP_028703829.1 | 483 | 141 | |||
XM_028847998.1 | XP_028703831.1 | 483 | 141 | |||
GenBank mRNA | JU322009.1 | - | 431 | 141 | ||
JU473802.1 | - | 470 | 141 | |||
JV047100.1 | - | 350 | 141 | |||
KC734514.1 | - | 239 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |