Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257439.1 | NP_001244368.1 | 312 | 75 | ||
XM_015142336.2 | XP_014997822.1 | 481 | 75 | |||
XM_015142351.2 | XP_014997837.1 | 377 | 75 | |||
GenBank mRNA | JU332909.1 | - | 312 | 75 | ||
JU474305.1 | - | 256 | 75 | |||
JV046184.1 | - | 355 | 75 | |||
JV642179.1 | - | 302 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |