Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258083.1 | NP_001245012.1 | 637 | 127 | ||
GenBank mRNA | JU326905.1 | - | 446 | 127 | ||
JU475276.1 | - | 637 | 127 | |||
JV046839.1 | - | 691 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |